Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124213.5 | NP_568696.1 | 2 - 3 | 1321 | 143 | |
GenBank mRNA | AF367312.1 | - | 2 - 3 | 1081 | 143 | |
AY074386.1 | - | 2 - 3 | 1069 | 143 | ||
FJ708793.1 | - | 1 - 2 | 835 | 143 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |