Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GKN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001148.1 | NP_001001148.1 | 2 - 3 | 103 | 80 | |
GenBank mRNA | AY456959.1 | - | 2 - 3 | 103 | 80 |
Set Membership: |
![]() ![]() ![]() ![]() |