Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076025.2 | NP_001069493.2 | 1 - 2 | 160 | 80 | |
GenBank mRNA | BC149334.1 | - | 1 - 2 | 184 | 80 | |
BT026194.1 | - | 168 | 80 | |||
DV887318.1 | - | 1 - 2 | 160 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |