Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206738.1 | NP_001193667.1 | 8 - 9 | 1258 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |