Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078088.3 | NP_510489.1 | 1 - 2 | 94 | 123 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |