Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078089.5 | NP_510490.1 | 14 - 15 | 2619 | 66 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |