Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: msh-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058801.7 | NP_491202.1 | 8 - 9 | 2545 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |