Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069586.8 | NP_501987.1 | 2 - 3 | 118 | 80 | |
GenBank mRNA | U20903.1 | - | 1 - 2 | 89 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |