Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069710.6 | NP_502111.3 | 21 - 22 | 3389 | 152 | |
GenBank mRNA | AY726541.1 | - | 21 - 22 | 3319 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |