Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pfn-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075804.6 | NP_508205.1 | 3 - 4 | 316 | 83 | |
GenBank mRNA | AY530910.1 | - | 3 - 4 | 288 | 83 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |