Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alh-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077731.7 | NP_510132.2 | 4 - 5 | 476 | 77 | |
NM_077732.8 | NP_510133.2 | 4 - 5 | 468 | 77 |
Set Membership: |
![]() ![]() ![]() ![]() |