Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083303.5 | NP_001076772.1 | 11 - 12 | 1439 | 87 |
Set Membership: |
![]() ![]() ![]() |