Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cope | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246677.1 | NP_001233606.1 | 8 - 9 | 843 | 84 | |
GenBank mRNA | JI891514.1 | - | 8 - 9 | 850 | 84 | |
JP045922.1 | - | 8 - 9 | 850 | 84 | ||
Z32554.1 | - | 8 - 9 | 843 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |