Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35g6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497182.2 | XP_003497230.2 | 1 - 2 | 449 | 75 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |