Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fitm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502066.2 | XP_003502114.1 | 1 - 2 | 261 | 91 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |