Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3hav1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643836.1 | XP_007642026.1 | 3 - 4 | 451 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |