Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646474.1 | XP_007644664.1 | 1 | 722 | 72 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |