Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285754.1 | NP_001272683.1 | 6 - 7 | 442 | 105 | |
GenBank mRNA | JN107562.1 | - | 6 - 7 | 442 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Capra hircus Probe spans exons |