Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013963641.2 | XP_013819095.2 | 8 - 9 | 744 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |