Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684015.3 | XP_005684072.1 | 3 - 4 | 859 | 79 | |
GenBank mRNA | JT631086.1 | - | 6 | 87 |
Set Membership: |
Amplicon length between 71 and 85 Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |