Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lectin-33A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103675.2 | NP_001097145.1 | 2 | 495 | 68 |
Set Membership: |
![]() ![]() ![]() ![]() |