Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JhI-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259067.1 | NP_001245996.1 | 3 - 4 | 580 | 71 | |
NM_001298934.1 | NP_001285863.1 | 3 - 4 | 550 | 71 | ||
NM_001298935.1 | NP_001285864.1 | 3 - 4 | 613 | 71 | ||
NM_135698.4 | NP_609542.1 | 3 - 4 | 556 | 71 | ||
NM_164988.4 | NP_723717.2 | 3 - 4 | 520 | 71 | ||
GenBank mRNA | AF273478.1 | - | 3 - 4 | 518 | 71 | |
AY051907.1 | - | 3 - 4 | 505 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |