Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299680.1 | NP_001286609.1 | 1 - 2 | 703 | 70 | |
NM_166339.2 | NP_725867.1 | 1 - 2 | 337 | 70 | ||
GenBank mRNA | BT023269.1 | - | 1 - 2 | 319 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() |