Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136953.4 | NP_610797.1 | 2 - 3 | 324 | 89 | |
GenBank mRNA | AY071739.1 | - | 2 - 3 | 345 | 89 |
Set Membership: |
![]() ![]() ![]() |