Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079177.4 | NP_523901.1 | 1 | 323 | 95 | |
GenBank mRNA | AY119009.1 | - | 1 | 310 | 95 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |