Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xe7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169093.3 | NP_730984.1 | 6 - 7 | 2325 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |