Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169364.3 | NP_731510.1 | 2 - 3 | 462 | 77 | |
GenBank mRNA | AY069252.1 | - | 2 - 3 | 417 | 77 |
Set Membership: |
![]() ![]() ![]() |