Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gukh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104364.2 | NP_001097834.2 | 4 - 5 | 2067 | 72 | |
NM_169845.2 | NP_732393.1 | 4 - 5 | 2067 | 72 |
Set Membership: |
![]() ![]() ![]() |