Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cyt-c-p | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299055.1 | NP_001285984.1 | 1 | 242 | 95 | |
NM_057828.4 | NP_477176.1 | 2 | 289 | 95 | ||
GenBank mRNA | AY071701.1 | - | 2 | 347 | 95 | |
KX531555.1 | - | 1 | 203 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |