Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obp84a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104230.3 | NP_001097700.2 | 2 - 3 | 376 | 149 | |
NM_057642.3 | NP_476990.1 | 1 - 2 | 257 | 149 | ||
GenBank mRNA | BT032807.1 | - | 1 - 2 | 238 | 149 | |
BT032823.1 | - | 1 - 2 | 238 | 149 | ||
U05984.1 | - | 1 - 2 | 258 | 149 |
Set Membership: |
![]() ![]() |