Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mlpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079679.3 | NP_001073147.1 | 12 - 13 | 1477 | 83 | |
XM_005168711.4 | XP_005168768.1 | 13 - 14 | 1689 | 83 | ||
XM_005168712.4 | XP_005168769.1 | 13 - 14 | 1646 | 83 | ||
XM_005168713.4 | XP_005168770.1 | 12 - 13 | 1518 | 83 | ||
GenBank mRNA | BC124539.1 | - | 11 - 12 | 1429 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |