Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl35a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002487.1 | NP_001002487.1 | 4 - 5 | 364 | 98 | |
GenBank mRNA | BC076321.1 | - | 4 - 5 | 364 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |