Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006510.1 | NP_001006510.1 | 9 - 10 | 1228 | 86 | |
GenBank mRNA | AB185935.1 | - | 9 - 10 | 1212 | 86 | |
AJ720056.1 | - | 9 - 10 | 1228 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |