Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC547956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249636.1 | NP_001236565.1 | 1 | 856 | 97 | |
GenBank mRNA | AF532621.1 | - | 1 | 856 | 97 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |