Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006756.hg.1 |
ENST00000320033.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006756.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006756.hg.1 |
TC17000551.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006755.hg.1 |
ENST00000320033.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006755.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006755.hg.1 |
TC17000551.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007949.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007949.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007949.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007949.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007949.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007949.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007949.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007949.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007949.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007949.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007949.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006757.hg.1 |
ENST00000320033.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006757.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006757.hg.1 |
TC17000551.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700158117.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700158117.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700158117.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700158117.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700158117.hg.1 |
TC1700007949.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700158118.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700158118.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700158118.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700158118.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700158118.hg.1 |
TC1700007949.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700158116.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700158116.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700158116.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700158116.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700158116.hg.1 |
TC1700007949.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700007949.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007949.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007949.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007949.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007949.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007949.hg.1 |
ENST00000320033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007949.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007949.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007949.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007949.hg.1 |
NM_001661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007949.hg.1 |
NM_001661.3 |
Refseq |