Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000511150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000511150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000511150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000511471.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000511471.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000511471.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000511471.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000511471.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000511471.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000511471.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000515146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000515146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000515146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000515146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000515146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
ENST00000515146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016167.hg.1 |
TC04001176.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016167.hg.1 |
ENST00000381501.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016167.hg.1 |
ENST00000511471.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016167.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000511150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000511150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000511150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000511471.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000511471.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000511471.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000511471.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000511471.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000511471.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000511471.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000515146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000515146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000515146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000515146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000515146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
ENST00000515146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010618.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172460.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172460.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172460.hg.1 |
ENST00000511471.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172460.hg.1 |
ENST00000515146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172460.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172460.hg.1 |
ENST00000381501.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172460.hg.1 |
ENST00000505452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172460.hg.1 |
ENST00000511471.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172460.hg.1 |
ENST00000515146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172460.hg.1 |
NM_003215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172460.hg.1 |
TC0400010618.hg.1 |
Clariom D TC |