Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024955.hg.1 |
TC05001828.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024955.hg.1 |
ENST00000274721.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024955.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024955.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024953.hg.1 |
TC05001828.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024953.hg.1 |
ENST00000274721.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024953.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024956.hg.1 |
TC05001828.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024956.hg.1 |
ENST00000274721.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024956.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024956.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024954.hg.1 |
TC05001828.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024954.hg.1 |
ENST00000274721.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024954.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024954.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184331.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184331.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184331.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184331.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184331.hg.1 |
TC0500012164.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012164.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184332.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184332.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184332.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184332.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184332.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184332.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184332.hg.1 |
TC0500012164.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500184333.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184333.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184333.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184333.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184333.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184333.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184333.hg.1 |
TC0500012164.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500184334.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184334.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184334.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184334.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184334.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184334.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184334.hg.1 |
TC0500012164.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500184335.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184335.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184335.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184335.hg.1 |
ENST00000274721.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184335.hg.1 |
ENST00000378362.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184335.hg.1 |
NM_001496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184335.hg.1 |
TC0500012164.hg.1 |
Clariom D TC |