Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000572959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000572959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000573815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000573815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000573815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000573815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000573815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000573815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000706.hg.1 |
ENST00000320785.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000706.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000706.hg.1 |
TC17000060.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000572959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000572959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000573815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000573815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000573815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000573815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000573815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000573815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
ENST00000574165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006660.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700145772.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145772.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700145772.hg.1 |
ENST00000320785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145772.hg.1 |
NM_006612.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700145772.hg.1 |
TC1700006660.hg.1 |
Clariom D TC |