Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019400.hg.1 |
TC07001245.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019400.hg.1 |
ENST00000222803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019400.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019399.hg.1 |
TC07001245.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019399.hg.1 |
ENST00000222803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019399.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019401.hg.1 |
TC07001245.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019401.hg.1 |
ENST00000222803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019401.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019396.hg.1 |
TC07001245.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019396.hg.1 |
ENST00000222803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07019396.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175502.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175502.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175502.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175502.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175502.hg.1 |
TC0700010621.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700175505.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175505.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175505.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175505.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175505.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175505.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175505.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175505.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175505.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175505.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175505.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175505.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175505.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175505.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175505.hg.1 |
TC0700010621.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700175507.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175507.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175507.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175507.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175507.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175507.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175507.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175507.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175507.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175507.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175507.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175507.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175507.hg.1 |
TC0700010621.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700175508.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175508.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175508.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175508.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175508.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175508.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175508.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175508.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175508.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175508.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175508.hg.1 |
TC0700010621.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010621.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175504.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175504.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175504.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175504.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175504.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175504.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175504.hg.1 |
ENST00000412494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175504.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175504.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175504.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175504.hg.1 |
TC0700010621.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700175506.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175506.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175506.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175506.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175506.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175506.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175506.hg.1 |
ENST00000222803.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175506.hg.1 |
ENST00000419018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175506.hg.1 |
ENST00000479939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700175506.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175506.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175506.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175506.hg.1 |
TC0700010621.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700175510.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175510.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175510.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175510.hg.1 |
NM_017946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175510.hg.1 |
NR_046478.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175510.hg.1 |
NR_046479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700175510.hg.1 |
TC0700010621.hg.1 |
Clariom D TC |