Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103187 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103199 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103190 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103185 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124198 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124194 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103201 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15041938 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103176 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103189 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103203 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103191 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103196 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103179 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103173 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103184 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124221 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103197 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103202 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103194 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124210 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103182 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20124190 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103195 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103198 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103200 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103178 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103192 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00103172 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
177512 |
AF087861.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
177512 |
AF165522.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
177512 |
AF217985.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
177512 |
AF498935.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
177512 |
AK291385.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
177512 |
BC033081.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
177512 |
BC046927.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
177512 |
NM_016154.3 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
150612 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
33682 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
26808 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
611139 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
492657 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P160800 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P160800 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
41262 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P200011 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P117463 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P117463 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
54039 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
34372 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P00033933 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_14_P200011 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P160800 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_23_P160800 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103197 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103202 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15041938 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124210 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124221 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103191 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103188 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103187 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103196 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103194 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103189 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103192 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103199 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124222 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103203 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103190 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124223 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103185 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103200 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103191 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103199 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15041938 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103203 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103189 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124222 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103188 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124221 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103192 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103196 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124223 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103190 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103187 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103202 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103197 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P20124210 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103194 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103200 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00103185 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_23_P160800 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P200011 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P117463 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103188 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103185 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103199 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103189 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103204 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103205 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103206 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103191 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103208 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103192 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103194 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103197 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P00103203 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00103208 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00103209 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P00103207 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P117463 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P200011 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P200011 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P117463 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P160800 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
19972 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
28410 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
80830 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
58058 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P200011 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P117463 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00033933 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103208 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103212 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15041938 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103199 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103179 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103204 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103184 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103188 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103197 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103219 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103214 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103205 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103191 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103175 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103152 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103210 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103192 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103185 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103216 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15041942 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103194 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103173 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103178 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103203 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103224 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103221 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103182 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00103176 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P117463 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P200011 |
- |
- |