Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000390185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000558632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000558632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000558632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000558632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000558632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
ENST00000558632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010733.hg.1 |
NR_030599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003266.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003266.hg.1 |
ENST00000396650.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003266.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003266.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003266.hg.1 |
TC15000357.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000390185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000558632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000558632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000558632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000558632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000558632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
ENST00000558632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010733.hg.1 |
NR_030599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182212.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182212.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182212.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182212.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182212.hg.1 |
ENST00000344300.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182212.hg.1 |
ENST00000396650.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182212.hg.1 |
NM_032413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182212.hg.1 |
NM_197955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182212.hg.1 |
TC1500010733.hg.1 |
Clariom D TC |