Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000525439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000525439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000525439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000525439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000525439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000534317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000534317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
ENST00000534317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002866.hg.1 |
ENST00000282096.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002866.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002866.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002866.hg.1 |
TC11000214.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000525439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000525439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000525439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000525439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000525439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000534317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000534317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
ENST00000534317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006923.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148713.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148713.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148713.hg.1 |
ENST00000525439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148713.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148713.hg.1 |
ENST00000282096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148713.hg.1 |
ENST00000455098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148713.hg.1 |
ENST00000525439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100148713.hg.1 |
NM_000922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100148713.hg.1 |
TC1100006923.hg.1 |
Clariom D TC |