Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035833.hg.1 |
TC03002005.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035833.hg.1 |
ENST00000241256.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035833.hg.1 |
ENST00000427970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035833.hg.1 |
NM_004122.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035833.hg.1 |
NM_198407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035834.hg.1 |
TC03002005.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035834.hg.1 |
ENST00000241256.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035834.hg.1 |
ENST00000427970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035834.hg.1 |
NM_004122.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035834.hg.1 |
NM_198407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013143.hg.1 |
ENST00000241256.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013143.hg.1 |
ENST00000241256.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013143.hg.1 |
ENST00000241256.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013143.hg.1 |
ENST00000241256.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013143.hg.1 |
ENST00000427970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013143.hg.1 |
ENST00000427970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013143.hg.1 |
ENST00000427970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013143.hg.1 |
NM_004122.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013143.hg.1 |
NM_004122.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013143.hg.1 |
NM_004122.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013143.hg.1 |
NM_198407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013143.hg.1 |
NM_198407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013143.hg.1 |
NM_198407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013143.hg.1 |
NM_198407.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013143.hg.1 |
NM_198407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198227.hg.1 |
ENST00000241256.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198227.hg.1 |
ENST00000427970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198227.hg.1 |
NM_004122.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198227.hg.1 |
NM_198407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198227.hg.1 |
ENST00000241256.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198227.hg.1 |
ENST00000427970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198227.hg.1 |
NM_004122.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198227.hg.1 |
NM_198407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198227.hg.1 |
TC0300013143.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300013143.hg.1 |
ENST00000241256.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013143.hg.1 |
ENST00000241256.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013143.hg.1 |
ENST00000241256.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013143.hg.1 |
ENST00000241256.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013143.hg.1 |
ENST00000427970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013143.hg.1 |
ENST00000427970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013143.hg.1 |
ENST00000427970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013143.hg.1 |
NM_004122.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013143.hg.1 |
NM_004122.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013143.hg.1 |
NM_004122.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013143.hg.1 |
NM_198407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013143.hg.1 |
NM_198407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013143.hg.1 |
NM_198407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013143.hg.1 |
NM_198407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013143.hg.1 |
NM_198407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198228.hg.1 |
ENST00000241256.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198228.hg.1 |
ENST00000427970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198228.hg.1 |
NM_004122.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198228.hg.1 |
NM_198407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198228.hg.1 |
ENST00000241256.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198228.hg.1 |
ENST00000427970.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198228.hg.1 |
NM_004122.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198228.hg.1 |
NM_198407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198228.hg.1 |
TC0300013143.hg.1 |
Clariom D TC |