Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H3J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003535.2 | NP_003526.1 | 1 | 428 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Human Interferon Pathway 1700 Inventoried Human_Interferon_Pathway Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630347 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630362 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820558 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630358 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630350 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630348 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630367 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630359 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630357 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630355 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820568 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820559 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820573 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820550 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630363 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820572 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 45178 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 6651 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P93282 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P93282 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 7688 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P105270 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P105270 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 21205 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P93282 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P93282 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820559 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630347 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630355 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820558 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820554 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820573 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820550 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630367 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630363 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630357 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820560 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820568 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630359 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820572 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820556 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630362 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630350 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820558 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820559 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820560 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630357 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820556 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630347 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820554 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820573 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630350 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630355 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820572 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630362 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630359 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820550 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820568 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630363 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630367 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P93282 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P105270 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630349 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630352 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630348 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630361 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630357 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630366 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630364 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630362 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630354 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630359 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630367 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630346 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04630360 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04630359 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04630361 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04630358 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15796574 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P105270 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P105270 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P93282 | - | - |
Custom Human Agilent Array 11k, Chromosome 6, Expression | 5070 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 4221 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 3392 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P105270 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630379 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630345 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630348 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630399 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630361 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630377 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630396 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630398 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630351 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630394 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630370 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630368 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630384 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630392 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630385 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630401 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630366 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630374 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630381 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630389 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630354 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630405 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630364 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630357 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630352 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630359 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630383 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630403 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P105270 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_003535_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8124538 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8124539 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8124537 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 214646_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11741809_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11741809_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR06016369.hg.1 | TC06001429.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06016369.hg.1 | ENST00000359303.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06016369.hg.1 | ENST00000479986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06016369.hg.1 | NM_003535.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011235.hg.1 | ENST00000359303.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011235.hg.1 | ENST00000359303.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011235.hg.1 | ENST00000359611.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011235.hg.1 | ENST00000359611.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011235.hg.1 | ENST00000479986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011235.hg.1 | ENST00000479986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011235.hg.1 | NM_003514.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011235.hg.1 | NM_003514.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011235.hg.1 | NM_003514.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011235.hg.1 | NM_003514.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011235.hg.1 | NM_003535.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011235.hg.1 | NM_003535.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011235.hg.1 | NM_003535.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06016368.hg.1 | TC06001429.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06016368.hg.1 | ENST00000359303.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06016368.hg.1 | NM_003535.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06016367.hg.1 | TC06001429.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06016367.hg.1 | ENST00000359303.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06016367.hg.1 | NM_003535.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600175744.hg.1 | ENST00000359303.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600175744.hg.1 | ENST00000479986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600175744.hg.1 | NM_003535.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600175744.hg.1 | ENST00000359303.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600175744.hg.1 | ENST00000479986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600175744.hg.1 | NM_003535.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600175744.hg.1 | TC0600011235.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600175742.hg.1 | NM_003535.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600175742.hg.1 | NM_003535.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600175742.hg.1 | TC0600011235.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600175743.hg.1 | ENST00000359303.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600175743.hg.1 | NM_003535.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600175743.hg.1 | ENST00000359303.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600175743.hg.1 | NM_003535.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600175743.hg.1 | TC0600011235.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600011235.hg.1 | ENST00000359303.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011235.hg.1 | ENST00000359303.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011235.hg.1 | ENST00000359611.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011235.hg.1 | ENST00000359611.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011235.hg.1 | ENST00000479986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011235.hg.1 | ENST00000479986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011235.hg.1 | NM_003514.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011235.hg.1 | NM_003514.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011235.hg.1 | NM_003514.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011235.hg.1 | NM_003514.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011235.hg.1 | NM_003535.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011235.hg.1 | NM_003535.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011235.hg.1 | NM_003535.2 | Refseq |
Human Genome U133 Plus 2.0 Array | 208575_at | - | - |
Human Genome U133 Plus 2.0 Array | 208577_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 208496_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 208496_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 214616_at | - | - |
Human Genome U133 Plus 2.0 Array | 208576_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 214616_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 208506_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 208506_at | - | - |
Human Genome U133 Plus 2.0 Array | 214472_at | - | - |
Human Genome U133 Plus 2.0 Array | 208575_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 214646_at | - | - |
Human Genome U133 Plus 2.0 Array | 208576_PM_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 208577_at | - | - |
Human Genome U133 Plus 2.0 Array | 214472_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 214646_PM_at | - | - |
Human Genome U133A 2.0 Array | 214472_PM_at | - | - |
Human Genome U133A 2.0 Array | 214616_PM_at | - | - |
Human Genome U133A 2.0 Array | 208506_PM_at | - | - |
Human Genome U133A 2.0 Array | 208576_s_at | - | - |
Human Genome U133A 2.0 Array | 214616_at | - | - |
Human Genome U133A 2.0 Array | 208576_PM_s_at | - | - |
Human Genome U133A 2.0 Array | 208575_PM_at | - | - |
Human Genome U133A 2.0 Array | 214646_at | - | - |
Human Genome U133A 2.0 Array | 214472_at | - | - |
Human Genome U133A 2.0 Array | 208577_at | - | - |
Human Genome U133A 2.0 Array | 208575_at | - | - |
Human Genome U133A 2.0 Array | 208496_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 208577_PM_at | - | - |
Human Genome U133A 2.0 Array | 214646_PM_at | - | - |
Human Genome U133A 2.0 Array | 208496_x_at | - | - |
Human Genome U133A 2.0 Array | 208506_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw590555 | - | - |
Human Genome U95 Set | 35598_at | - | - |
Human Genome U95 Set | 35598_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214646_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214646_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214646_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214646_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214646_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214646_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214646_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214646_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214646_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214646_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214646_at9 | - | - |
Human Genome U133 Set | 208496_PM_x_at | - | - |
Human Genome U133 Set | 208575_PM_at | - | - |
Human Genome U133 Set | 208577_PM_at | - | - |
Human Genome U133 Set | 208577_at | - | - |
Human Genome U133 Set | 208576_s_at | - | - |
Human Genome U133 Set | 208506_PM_at | - | - |
Human Genome U133 Set | 208496_x_at | - | - |
Human Genome U133 Set | 214616_at | - | - |
Human Genome U133 Set | 208575_at | - | - |
Human Genome U133 Set | 214646_at | - | - |
Human Genome U133 Set | 208506_at | - | - |
Human Genome U133 Set | 214616_PM_at | - | - |
Human Genome U133 Set | 214472_PM_at | - | - |
Human Genome U133 Set | 214646_PM_at | - | - |
Human Genome U133 Set | 214472_at | - | - |
Human Genome U133 Set | 208576_PM_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029576 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029576 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 197470 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1707257 | NM_003535.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1707257 | NM_003535.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1707257 | NM_003535.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1707257 | NM_003535.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1707257 | NM_003535.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_15718724-S | NM_003535.2 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_29231-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0002370435 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0002370435 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1707257 | NM_003535.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1707257 | NM_003535.2 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 7650086 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_29231 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1707257 | NM_003535.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1707257 | NM_003535.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1707257 | NM_003535.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1707257 | NM_003535.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1707257 | NM_003535.2 | human_transcriptome |