Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000461829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000461829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000461829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000488482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000488482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000488482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000488482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000488482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000488482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
ENST00000507624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000307534.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000322191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000342512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000461829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000461829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000461829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000488482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000488482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000488482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000488482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000488482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000488482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
ENST00000507624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_138316.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011382.hg.1 |
NM_148978.2 |
Refseq |