Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002561.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002561.hg.1 |
ENST00000375749.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002561.hg.1 |
ENST00000486717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002561.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002561.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002561.hg.1 |
TC20000208.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000408740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000434904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000434904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000465791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000465791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000465791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000465791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000486717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000486717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000486717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000486717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000486717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000486717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
ENST00000486717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007110.hg.1 |
NR_031726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000408740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000434904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000434904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000465791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000465791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000465791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000465791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000486717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000486717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000486717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000486717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000486717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000486717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
ENST00000486717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007110.hg.1 |
NR_031726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148583.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148583.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148583.hg.1 |
ENST00000486717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148583.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148583.hg.1 |
NM_172236.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148583.hg.1 |
TC2000007110.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148583.hg.1 |
ENST00000375730.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148583.hg.1 |
ENST00000375749.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148583.hg.1 |
ENST00000486717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148583.hg.1 |
NM_015352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148583.hg.1 |
NM_172236.1 |
Refseq |