Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016358.hg.1 |
TC05001137.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016358.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016358.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000503112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000503112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000503112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000503112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000504058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000504058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000504058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000504058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
ENST00000504058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009987.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170288.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170288.hg.1 |
ENST00000503112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170288.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170288.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170288.hg.1 |
ENST00000503112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170288.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170288.hg.1 |
TC0500009987.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500170287.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170287.hg.1 |
ENST00000503112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170287.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170287.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170287.hg.1 |
ENST00000503112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170287.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170287.hg.1 |
TC0500009987.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000255764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000503112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000503112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000503112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000503112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000504058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000504058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000504058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000504058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
ENST00000504058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
NM_032286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009987.hg.1 |
NM_032286.2 |
Refseq |