Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009405 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009365 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752914 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752862 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009393 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009441 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752845 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009351 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752913 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752933 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752843 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009353 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009400 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009415 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009466 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009430 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752891 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752873 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009368 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009442 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752871 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009375 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009358 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009402 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009404 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009427 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752842 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009443 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009461 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752900 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752866 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752872 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009398 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009468 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009356 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752926 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752869 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009454 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009359 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752915 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009448 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009447 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009412 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16012774 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009424 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752921 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752920 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009434 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009387 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752896 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009352 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009397 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009381 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009384 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752938 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16012763 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009458 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009426 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009401 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009382 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752919 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009361 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009354 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752894 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009373 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009394 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009355 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752937 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009418 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009374 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009406 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009425 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009362 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009380 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009408 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009453 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752893 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009431 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009392 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752834 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009416 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009456 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752881 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009420 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752924 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009432 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009446 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009377 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752908 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009396 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009369 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009389 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009436 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752852 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009371 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752907 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05752867 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P26009462 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
2361 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
38549 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
492864 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3395547 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_3848 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_3849 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P124472 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P124472 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
35081 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P18100536 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3395547 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_33_P3395547 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_19_P00323890 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009365 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009400 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009380 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752852 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752869 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009404 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009375 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009374 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009359 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009361 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752894 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009360 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009408 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009396 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752862 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009369 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752866 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009384 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009368 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009371 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752896 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009381 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009373 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009412 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009402 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009387 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009405 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009401 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009393 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009397 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752867 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752873 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009389 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009410 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009382 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752845 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009362 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009398 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009377 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752842 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752843 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009360 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752843 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009396 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009361 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009375 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009405 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009381 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009410 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009368 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752869 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009380 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009359 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009397 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009387 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752896 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752867 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752852 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009377 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009374 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009398 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752842 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752873 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752862 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009400 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009384 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009382 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009393 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009365 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009412 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752866 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009401 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752894 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009369 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009373 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009404 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009362 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05752845 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009402 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009408 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009389 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P26009371 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P124472 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P124472 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P124472 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
170532 |
NM_006566.1 |
refseq_mrna_human_NM |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
170532 |
U56102.1 |
genbank_mrna_human |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
3419 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P18100536 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P38187076 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P25750368 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752927 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752869 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752924 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752876 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752895 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752909 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752905 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752910 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752874 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16012767 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752912 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752934 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752845 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752893 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752933 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752837 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752836 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752868 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752921 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752919 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752926 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752879 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16012771 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752940 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752886 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752848 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752862 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752872 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752834 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752882 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752931 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752897 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16012773 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752860 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752865 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752929 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752916 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752850 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16012777 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752911 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752906 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752923 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752918 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752890 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752938 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752914 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752856 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752841 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752915 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16012775 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752901 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752853 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752839 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752936 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05752843 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P18100536 |
- |
- |