Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03029573.hg.1 |
TC03001637.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03029573.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03029573.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015268.hg.1 |
ENST00000426321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015268.hg.1 |
ENST00000426321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009407.hg.1 |
ENST00000422272.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03029574.hg.1 |
TC03001637.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03029574.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03029574.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009407.hg.1 |
ENST00000422272.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189029.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189029.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300189029.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189029.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300189029.hg.1 |
TC0300011955.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011955.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015268.hg.1 |
ENST00000426321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015268.hg.1 |
ENST00000426321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189030.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189030.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300189030.hg.1 |
ENST00000478945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300189030.hg.1 |
NM_138815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300189030.hg.1 |
TC0300011955.hg.1 |
Clariom D TC |