Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN4IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318746.1 | NP_001305675.1 | 4 | 457 | 91 | |
NM_032730.5 | NP_116119.2 | 4 | 1043 | 91 | ||
XM_011536192.2 | XP_011534494.1 | 5 | 1131 | 91 | ||
XM_017011375.1 | XP_016866864.1 | 4 | 1043 | 91 | ||
XM_017011376.1 | XP_016866865.1 | 4 | 1043 | 91 | ||
GenBank mRNA | AF336861.1 | - | 4 | 617 | 91 | |
AF439711.1 | - | 4 | 566 | 91 | ||
AK095207.1 | - | 4 | 457 | 91 | ||
AK313962.1 | - | 4 | 961 | 91 | ||
AY063761.1 | - | 4 | 566 | 91 | ||
BC006399.2 | - | 4 | 838 | 4309263 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon |